Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550555.2 | XP_005550612.1 | 4 - 5 | 744 | 117 | |
XM_015447956.1 | XP_015303442.1 | 3 - 4 | 526 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |