Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582474.2 | XP_005582531.2 | 1 - 2 | 1161 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |