Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGLON5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590090.2 | XP_005590147.1 | 9 - 10 | 1964 | 62 | |
XM_015441679.1 | XP_015297165.1 | 10 - 11 | 2083 | 62 | ||
XM_015441680.1 | XP_015297166.1 | 10 - 11 | 2050 | 62 | ||
XM_015441681.1 | XP_015297167.1 | 10 - 11 | 2021 | 62 | ||
XM_015441682.1 | XP_015297168.1 | 10 - 11 | 2087 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |