Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285354.1 | NP_001272283.1 | 6 - 7 | 1557 | 57 | |
XM_005557474.2 | XP_005557531.1 | 6 - 7 | 1531 | 57 | ||
XM_005557475.2 | XP_005557532.1 | 6 - 7 | 1454 | 57 | ||
XM_005557476.2 | XP_005557533.1 | 6 - 7 | 1300 | 57 | ||
XM_015451390.1 | XP_015306876.1 | 5 - 6 | 1149 | 57 | ||
GenBank mRNA | AB168797.1 | - | 6 - 7 | 1557 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |