Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549946.2 | XP_005550003.1 | 4 - 5 | 474 | 83 | |
XM_005549947.2 | XP_005550004.1 | 3 - 4 | 422 | 83 | ||
XM_015447621.1 | XP_015303107.1 | 4 - 5 | 500 | 83 | ||
XM_015447622.1 | XP_015303108.1 | 4 - 5 | 982 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |