Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557353.2 | XP_005557410.1 | 5 - 6 | 551 | 75 | |
XM_005557354.2 | XP_005557411.1 | 5 - 6 | 513 | 75 | ||
XM_005557355.2 | XP_005557412.1 | 5 - 6 | 510 | 75 | ||
XM_005557356.2 | XP_005557413.1 | 5 - 6 | 496 | 75 | ||
XM_005557357.2 | XP_005557414.1 | 5 - 6 | 524 | 75 | ||
XM_005557358.2 | XP_005557415.1 | 5 - 6 | 468 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |