Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284543.1 | NP_001271472.1 | 3 - 4 | 317 | 75 | |
GenBank mRNA | JQ288995.1 | - | 3 - 4 | 317 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |