Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572374.2 | XP_005572431.1 | 2 - 3 | 1084 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |