Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1H1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541058.2 | XP_005541115.1 | 6 - 7 | 819 | 56 | |
XM_015450853.1 | XP_015306339.1 | 6 - 7 | 856 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |