Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHAL6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578441.2 | XP_005578498.1 | 1 - 2 | 293 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |