Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283571.1 | NP_001270500.1 | 4 - 5 | 636 | 102 | |
XM_005541976.2 | XP_005542033.1 | 5 - 6 | 764 | 102 | ||
GenBank mRNA | AB171843.1 | - | 4 - 5 | 636 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |