Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015455091.1 | XP_015310577.1 | 7 - 8 | 1005 | 70 | |
GenBank mRNA | AB168988.1 | - | 7 - 8 | 810 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |