Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286147.1 | NP_001273076.1 | 2 - 3 | 226 | 129 | |
XM_015436506.1 | XP_015291992.1 | 2 - 3 | 228 | 129 | ||
XM_015436507.1 | XP_015291993.1 | 2 - 3 | 389 | 129 | ||
GenBank mRNA | AB174101.1 | - | 2 - 3 | 226 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |