Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580702.2 | XP_005580759.1 | 2 - 3 | 238 | 62 | |
XM_005580703.2 | XP_005580760.1 | 2 - 3 | 453 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |