Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581843.1 | XP_005581900.1 | 1 - 2 | 479 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |