Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546257.2 | XP_005546314.1 | 1 | 673 | 105 | |
XM_005546258.2 | XP_005546315.1 | 2 | 487 | 105 | ||
XM_005546259.2 | XP_005546316.1 | 2 | 476 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |