Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587811.2 | XP_005587868.1 | 2 - 3 | 236 | 79 | |
XM_005587812.2 | XP_005587869.1 | 2 - 3 | 289 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |