Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580128.2 | XP_005580185.1 | 2 - 3 | 469 | 139 | |
XM_005580130.2 | XP_005580187.1 | 1 - 2 | 133 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |