Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549284.2 | XP_005549341.1 | 2 - 3 | 426 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Inventoried Probe spans exons |