Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285270.1 | NP_001272199.1 | 2 - 3 | 319 | 128 | |
XM_005583490.2 | XP_005583547.1 | 2 - 3 | 331 | 128 | ||
GenBank mRNA | AB168602.1 | - | 2 - 3 | 319 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |