Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015450851.1 | XP_015306337.1 | 1 - 2 | 277 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |