Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577238.2 | XP_005577295.1 | 1 - 2 | 446 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |