Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556533.2 | XP_005556590.1 | 2 - 3 | 587 | 108 | |
XM_005556535.1 | XP_005556592.1 | 2 - 3 | 342 | 108 | ||
XM_005556536.2 | XP_005556593.2 | 2 - 3 | 804 | 108 | ||
XM_015451173.1 | XP_015306659.1 | 3 - 4 | 464 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |