Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIF1AN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566198.2 | XP_005566255.1 | 3 - 4 | 678 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Inventoried Probe spans exons |