Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579424.2 | XP_005579481.1 | 5 - 6 | 1556 | 134 | |
XM_005579425.2 | XP_005579482.1 | 5 - 6 | 1209 | 134 | ||
XM_015435627.1 | XP_015291113.1 | 4 - 5 | 3310 | 134 | ||
XM_015435628.1 | XP_015291114.1 | 5 - 6 | 2017 | 134 | ||
XM_015435629.1 | XP_015291115.1 | 5 - 6 | 1553 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |