Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005554843.2 | XP_005554900.1 | 2 - 3 | 416 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |