Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590141.2 | XP_005590198.1 | 2 | 1349 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Both primers and probe map within a single exon |