Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015457034.1 | XP_015312520.1 | 1 | 4618 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |