Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572952.2 | XP_005573009.1 | 1 - 2 | 1465 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |