Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582822.2 | XP_005582879.1 | 2 - 3 | 198 | 90 | |
XM_005582823.2 | XP_005582880.1 | 2 - 3 | 198 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |