Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582747.2 | XP_005582804.2 | 2 - 3 | 266 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |