Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575958.2 | XP_005576015.1 | 3 - 4 | 837 | 102 | |
XM_015433696.1 | XP_015289182.1 | 5 - 6 | 2018 | 102 | ||
XM_015433697.1 | XP_015289183.1 | 4 - 5 | 1923 | 102 | ||
XM_015433698.1 | XP_015289184.1 | 4 - 5 | 1748 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |