Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594364.2 | XP_005594421.1 | 3 - 4 | 6392 | 63 | |
XM_015443884.1 | XP_015299370.1 | 3 - 4 | 6430 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |