Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553564.2 | XP_005553621.1 | 2 | 530 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |