Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285236.1 | NP_001272165.1 | 11 - 12 | 1430 | 76 | |
XM_005593053.2 | XP_005593110.1 | 11 - 12 | 1578 | 76 | ||
GenBank mRNA | AB179464.1 | - | 11 - 12 | 1430 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |