Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563554.1 | XP_005563611.1 | 2 - 3 | 444 | 111 | |
GenBank mRNA | AB070008.1 | - | 4 - 5 | 476 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |