Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283390.1 | NP_001270319.1 | 1 - 2 | 263 | 68 | |
XM_015437384.1 | XP_015292870.1 | 1 - 2 | 825 | 68 | ||
GenBank mRNA | AB170621.1 | - | 1 - 2 | 263 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |