Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283669.1 | NP_001270598.1 | 2 - 3 | 88 | 109 | |
XM_005541168.2 | XP_005541225.1 | 2 - 3 | 154 | 109 | ||
XM_015452322.1 | XP_015307808.1 | 2 - 3 | 154 | 109 | ||
GenBank mRNA | AF485813.1 | - | 2 - 3 | 88 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |