Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284546.1 | NP_001271475.1 | 1 | 1325 | 87 | |
GenBank mRNA | AB070158.1 | - | 1 | 1325 | 87 | |
AB070198.1 | - | 1 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |