Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283542.1 | NP_001270471.1 | 2 - 3 | 208 | 101 | |
GenBank mRNA | AB169707.1 | - | 2 - 3 | 208 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |