Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283542.1 | NP_001270471.1 | 5 - 6 | 542 | 103 | |
GenBank mRNA | AB169707.1 | - | 5 - 6 | 542 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |