Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285289.1 | NP_001272218.1 | 3 - 4 | 392 | 107 | |
XM_005558118.2 | XP_005558175.1 | 3 - 4 | 742 | 107 | ||
GenBank mRNA | AB169519.1 | - | 3 - 4 | 432 | 107 | |
AB171857.1 | - | 3 - 4 | 392 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |