Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283676.1 | NP_001270605.1 | 4 - 5 | 597 | 91 | |
XM_005547965.2 | XP_005548022.1 | 4 - 5 | 518 | 91 | ||
XM_005547967.2 | XP_005548024.1 | 4 - 5 | 563 | 91 | ||
XM_015445279.1 | XP_015300765.1 | 4 - 5 | 492 | 91 | ||
GenBank mRNA | AB168592.1 | - | 4 - 5 | 600 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |