Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283364.1 | NP_001270293.1 | 4 - 5 | 837 | 100 | |
GenBank mRNA | AB174277.1 | - | 4 - 5 | 837 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |