Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284066.1 | NP_001270995.1 | 3 - 4 | 446 | 83 | |
XM_005583485.2 | XP_005583542.1 | 3 - 4 | 483 | 83 | ||
GenBank mRNA | AB168985.1 | - | 3 - 4 | 446 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |