Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284177.1 | NP_001271106.1 | 2 - 3 | 292 | 97 | |
XM_015430169.1 | XP_015285655.1 | 3 - 4 | 1451 | 97 | ||
XM_015430170.1 | XP_015285656.1 | 4 - 5 | 1894 | 97 | ||
XM_015430171.1 | XP_015285657.1 | 3 - 4 | 1451 | 97 | ||
XM_015430172.1 | XP_015285658.1 | 4 - 5 | 1894 | 97 | ||
GenBank mRNA | AJ585528.1 | - | 2 - 3 | 292 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |