Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287667.1 | NP_001274596.1 | 8 - 9 | 813 | 84 | |
XM_005574280.2 | XP_005574337.1 | 9 - 10 | 1029 | 84 | ||
GenBank mRNA | AB071068.1 | - | 8 - 9 | 813 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |