Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449302.1 | XP_015304788.1 | 40 - 41 | 8493 | 61 | |
XM_015449307.1 | XP_015304793.1 | 41 - 42 | 8412 | 61 | ||
XM_015449309.1 | XP_015304795.1 | 39 - 40 | 8351 | 61 | ||
XM_015449313.1 | XP_015304799.1 | 33 - 34 | 6329 | 61 | ||
XM_015449318.1 | XP_015304804.1 | 33 - 34 | 7548 | 61 | ||
XM_015449320.1 | XP_015304806.1 | 33 - 34 | 6330 | 61 | ||
XM_015449323.1 | XP_015304809.1 | 32 - 33 | 7135 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Inventoried Probe spans exons |