Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546276.2 | XP_005546333.1 | 2 - 3 | 504 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |