Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552510.2 | XP_005552567.1 | 3 - 4 | 1453 | 96 | |
XM_005552511.2 | XP_005552568.1 | 3 - 4 | 809 | 96 | ||
XM_005552512.2 | XP_005552569.1 | 3 - 4 | 807 | 96 |